Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-[(Polyethylene glycol)]-N,N-ditetradecylacetamide 97% PEG2000

Supplier: AMBEED, INC
Warning

A1375214-10MG A1375214-5MG
77621-946EA 85.49 USD InStock
77621-946 77621-948
2-[(Polyethylene glycol)]-N,N-ditetradecylacetamide 97% PEG2000
2-[(Polyethylene glycol)]-N,N-ditetradecylacetamide
Precaución: for Research Use Only


Formula: (C₂H₄O)nC₃₁H₆₃NO₂
CAS Number: 1849616-42-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty