Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Nitro-1H-pyrazol-5-yl)ethanone ≥98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A137740-100MG A137740-1G
77683-207EA 80 USD InStock
77683-207 77683-208
1-(4-Nitro-1H-pyrazol-5-yl)ethanone ≥98%, Ambeed.Inc
1-(4-Nitro-1H-pyrazol-5-yl)ethanone
Precaución: For research use only.


Formula: C₅H₅N₃O₃
MW: 155.11 g/mol
MDL Number: MFCD26743745
CAS Number: 2228374-35-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty