Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,4,8-Tri-boc-1,4,8,11-tetraazacyclotetradecane ≥97%

Supplier: AMBEED, INC
Warning

A139036-100MG A139036-1G A139036-250MG
77685-187EA 25 USD InStock
77685-187 77685-188 77685-189
1,4,8-Tri-boc-1,4,8,11-tetraazacyclotetradecane ≥97%
1,4,8-Tri-boc-1,4,8,11-tetraazacyclotetradecane
Precaución: for Research Use Only


Formula: C₂₅H₄₈N₄O₆
MDL Number: MFCD09263315
CAS Number: 170161-27-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty