Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 5-iodopentanoate ≥98%

Supplier: AMBEED, INC
Danger

A1400105-100MG A1400105-250MG A1400105-5G
77622-034EA 22.66 USD InStock
77622-034 77622-036 77622-038
tert-Butyl 5-iodopentanoate ≥98%
tert-Butyl 5-iodopentanoate
Precaución: for Research Use Only


Formula: C₉H₁₇IO₂
MW: 284.1346 g/mol
MDL Number: MFCD28401357
CAS Number: 56198-37-9
UN: 2810
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty