Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

RTA-402 ≥98%

Supplier: AMBEED, INC
Warning

A142891-100MG A142891-250MG
77671-034EA 67 USD InStock
77671-034 77671-036
RTA-402 ≥98%
RTA-402
Precaución: for Research Use Only


Formula: C₃₂H₄₃NO₄
MDL Number: MFCD11983137
CAS Number: 218600-53-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty