Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Merck-5 ≥98%

Supplier: AMBEED, INC
Warning

A143706-10MG A143706-2MG A143706-5MG
77685-218EA 86 USD InStock
77685-218 77685-219 77685-220
Merck-5 ≥98%
Merck-5
Precaución: for Research Use Only


Formula: C₁₈H₁₆FN₃O
MDL Number: MFCD17019334
CAS Number: 457081-03-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty