Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

rel-(3aR,5s,6aS)-N-(3-Methoxy-1,2,4-thiadiazol-5-yl)-5-(methyl(7H-pyrrolo[2,3-d]pyrimidin-4-yl)amino)hexahydrocyclopenta[c]pyrrole-2(1H)-carboxamide ≥98%

Supplier: AMBEED, INC
Warning

A1443404-25MG A1443404-50MG
77622-116EA 346.08 USD InStock
77622-116 77622-118
rel-(3aR,5s,6aS)-N-(3-Methoxy-1,2,4-thiadiazol-5-yl)-5-(methyl(7H-pyrrolo[2,3-d]pyrimidin-4-yl)amino)hexahydrocyclopenta[c]pyrrole-2(1H)-carboxamide ≥98%
rel-(3aR,5s,6aS)-N-(3-Methoxy-1,2,4-thiadiazol-5-yl)-5-(methyl(7H-pyrrolo[2,3-d]pyrimidin-4-yl)amino)hexahydrocyclopenta[c]pyrrole-2(1H)-carboxamide
Precaución: for Research Use Only


Formula: C₁₈H₂₂N₈O₂S
MDL Number: MFCD32701909
CAS Number: 1445987-21-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty