Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Alizarin complexone indicator

Supplier: AMBEED, INC
Warning

A1456845-25G A1456845-5G
77622-178EA 99.91 USD InStock
77622-178 77622-180
Alizarin complexone indicator
Alizarin complexone
Precaución: for Research Use Only


Formula: C₁₉H₁₅NO₈
MW: 385.33 g/mol
Melting Pt: 180
MDL Number: MFCD00001202
CAS Number: 3952-78-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty