Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hyaluronic acid sodium salt, M.W. 8000-15000

Supplier: AMBEED, INC
Warning

A1463892-100G A1463892-25G
77597-330EA 62.53 USD InStock
77597-330 77597-332
Hyaluronic acid sodium salt, M.W. 8000-15000
Hyaluronic acid sodium salt
Precaución: for Research Use Only


Formula: (C₁₄H₂₁NaNO₁₁)n
MDL Number: MFCD00875848
CAS Number: 9067-32-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty