Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,6-Dichloro-N-(4'-fluoro-[1,1'-biphenyl]-4-yl)-1,3,5-triazin-2-amine ≥98%

Supplier: AMBEED, INC
Warning

A1465541-100MG A1465541-50MG
77624-236EA 211.15 USD InStock
77624-236 77624-238
4,6-Dichloro-N-(4'-fluoro-[1,1'-biphenyl]-4-yl)-1,3,5-triazin-2-amine ≥98%
4,6-Dichloro-N-(4'-fluoro-[1,1'-biphenyl]-4-yl)-1,3,5-triazin-2-amine
Precaución: for Research Use Only


Formula: C₁₅H₉Cl₂FN₄
MDL Number: MFCD31697744
CAS Number: 2138882-71-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty