Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-(5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl)acetamido)-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxamide ≥98%

Supplier: AMBEED, INC
Warning

A1465554-100MG A1465554-1G A1465554-250MG
77624-240EA 8.24 USD InStock
77624-240 77624-242 77624-244
2-(2-(5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl)acetamido)-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxamide ≥98%
2-(2-(5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl)acetamido)-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxamide
Precaución: for Research Use Only


Formula: C₁₆H₁₇F₃N₄O₂S
MW: 386.3920 g/mol
MDL Number: MFCD02053612
CAS Number: 489408-02-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty