Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-2H-indazole-5-boronic acid pinacol ester ≥98%

Supplier: AMBEED, INC
Warning

A1477078-250MG A1477078-1G A1477078-5G
77597-508EA 13.33 USD InStock
77597-508 77624-330 77624-332
2-Methyl-2H-indazole-5-boronic acid pinacol ester ≥98%
2-Methyl-2H-indazole-5-boronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₄H₁₉BN₂O₂
MDL Number: MFCD11109436
CAS Number: 1189746-27-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty