Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl oct-7-yn-1-ylcarbamate ≥95%

Supplier: AMBEED, INC
Warning

A148076-100MG A148076-1G A148076-250MG A148076-5G
77748-031EA 58 USD InStock
77748-031 77748-032 77748-033 77748-034
tert-Butyl oct-7-yn-1-ylcarbamate ≥95%
tert-Butyl oct-7-yn-1-ylcarbamate
Precaución: for Research Use Only


Formula: C₁₃H₂₃NO₂
CAS Number: 1451262-84-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty