Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2S,3R)-1-(3-Cyano-6-methyl-4-(trifluoromethyl)pyridin-2-yl)-3-hydroxy-N-methyl-N-(m-tolyl)pyrrolidine-2-carboxamide ≥98%

Supplier: AMBEED, INC

A1488545-1MG A1488545-10MG A1488545-5MG
77597-638EA 152.73 USD InStock
77597-638 77622-250 77622-252
(2S,3R)-1-(3-Cyano-6-methyl-4-(trifluoromethyl)pyridin-2-yl)-3-hydroxy-N-methyl-N-(m-tolyl)pyrrolidine-2-carboxamide ≥98%
(2S,3R)-1-(3-Cyano-6-methyl-4-(trifluoromethyl)pyridin-2-yl)-3-hydroxy-N-methyl-N-(m-tolyl)pyrrolidine-2-carboxamide
Precaución: for Research Use Only


Formula: C₂₁H₂₁F₃N₄O₂
CAS Number: 2603528-97-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty