Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Aminobutyl)-2-butyl-1H-imidazo[4,5-c]quinolin-4-amine ≥95%

Supplier: AMBEED, INC
Warning

A1504220-1G A1504220-250MG A1504220-50MG
77624-732EA 535.6 USD InStock
77624-732 77624-734 77624-736
1-(4-Aminobutyl)-2-butyl-1H-imidazo[4,5-c]quinolin-4-amine ≥95%
1-(4-Aminobutyl)-2-butyl-1H-imidazo[4,5-c]quinolin-4-amine
Precaución: for Research Use Only


Formula: C₁₈H₂₅N₅
MDL Number: MFCD33398409
CAS Number: 313350-31-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty