Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

UDP-Xylose disodium salt ≥98%

Supplier: AMBEED, INC
Warning

A1508162-10MG A1508162-250MG A1508162-100MG A1508162-50MG
77597-824EA 34.85 USD InStock
77597-824 77597-826 77624-786 77624-788
UDP-Xylose disodium salt ≥98%
UDP-Xylose disodium salt
Precaución: for Research Use Only


Formula: C₁₄H₂₂N₂Na₂O₁₆P₂
MW: 582.25 g/mol
MDL Number: MFCD15145136
CAS Number: 108320-89-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty