Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Chloro-4-(prop-2-yn-1-yloxy)benzene ≥97%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A151629-1G A151629-250MG
77683-278EA 71 USD InStock
77683-278 77683-279
1-Chloro-4-(prop-2-yn-1-yloxy)benzene ≥97%, Ambeed.Inc
1-Chloro-4-(prop-2-yn-1-yloxy)benzene
Precaución: For research use only.


Formula: C₉H₇ClO
MDL Number: MFCD09836692
CAS Number: 19130-39-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty