Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 4-(azetidin-3-yl)piperazine-1-carboxylate hydrochloride ≥98%

Supplier: AMBEED, INC
Warning

A151956-100MG A151956-10G A151956-5G
77667-140EA 23 USD InStock
77667-140 77667-142 77667-144
tert-Butyl 4-(azetidin-3-yl)piperazine-1-carboxylate hydrochloride ≥98%
tert-Butyl 4-(azetidin-3-yl)piperazine-1-carboxylate hydrochloride
Precaución: for Research Use Only


Formula: C₁₂H₂₄ClN₃O₂
MDL Number: MFCD17169879
CAS Number: 178312-58-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty