Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

rel-ethyl (1R,8S,9r)-bicyclo[6.1.0]non-4-ene-9-carboxylate ≥95%

Supplier: AMBEED, INC
Warning

A1520969-250MG A1520969-100MG
77625-200EA 159.65 USD InStock
77625-200 77625-198
rel-ethyl (1R,8S,9r)-bicyclo[6.1.0]non-4-ene-9-carboxylate ≥95%
rel-ethyl (1R,8S,9r)-bicyclo[6.1.0]non-4-ene-9-carboxylate
Precaución: for Research Use Only


Formula: C₁₂H₁₈O₂
MW: 194.27 g/mol
MDL Number: MFCD20264902
CAS Number: 79549-89-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty