Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dibromo(4,4'-dimethoxy-2,2'-bipyridine-κN1,κN1')Nickel ≥95%

Supplier: AMBEED, INC
Danger

A1543162-1G A1543162-250MG A1543162-5G
77625-354EA 122.57 USD InStock
77625-354 77625-356 77625-358
Dibromo(4,4'-dimethoxy-2,2'-bipyridine-κN1,κN1')Nickel ≥95%
Dibromo(4,4'-dimethoxy-2,2'-bipyridine-κN1,κN1')Nickel
Precaución: for Research Use Only


Formula: C₁₂H₁₂Br₂N₂NiO₂
CAS Number: 2409691-04-7
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty