Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-10-hydroxycamptothecin 98+%

Supplier: AMBEED, INC
Danger

A155153-5G A155153-1MG
77600-212EA 145.55 USD InStock
77600-212 77625-410
(S)-10-hydroxycamptothecin 98+%
(S)-10-hydroxycamptothecin
Precaución: for Research Use Only


Formula: C₂₀H₁₆N₂O₅
MW: 364.35 g/mol
MDL Number: MFCD02093100
CAS Number: 19685-09-7
UN: 1544
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty