Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(6-((2,5-Dioxopyrrolidin-1-yl)oxy)-6-oxohexyl)-3,3-dimethyl-2-(5-(1,3,3-trimethylindolin-2-ylidene)penta-1,3-dien-1-yl)-3H-indol-1-ium tetrafluoroborate ≥95%

Supplier: AMBEED, INC
Danger

A1560187-100MG A1560187-10MG A1560187-250MG
77625-422EA 655.08 USD InStock
77625-422 77625-424 77625-426
1-(6-((2,5-Dioxopyrrolidin-1-yl)oxy)-6-oxohexyl)-3,3-dimethyl-2-(5-(1,3,3-trimethylindolin-2-ylidene)penta-1,3-dien-1-yl)-3H-indol-1-ium tetrafluoroborate ≥95%
1-(6-((2,5-Dioxopyrrolidin-1-yl)oxy)-6-oxohexyl)-3,3-dimethyl-2-(5-(1,3,3-trimethylindolin-2-ylidene)penta-1,3-dien-1-yl)-3H-indol-1-ium tetrafluoroborate
Precaución: for Research Use Only


Formula: C₃₆H₄₂BF₄N₃O₄
MW: 667.54 g/mol
CAS Number: 1263093-76-0
UN: 1759
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty