Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ginkgolide C ≥98%

Supplier: AMBEED, INC
Warning

A156718-10MG A156718-1MG
77748-068EA 14 USD InStock
77748-068 77748-069
Ginkgolide C ≥98%
Ginkgolide C
Precaución: for Research Use Only


Formula: C₂₀H₂₄O₁₁
MW: 440.40 g/mol
MDL Number: MFCD02094178
CAS Number: 15291-76-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty