Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(-)-Huperzine A ≥98%

Supplier: AMBEED, INC
Danger

A161814-100MG A161814-250MG
77748-088EA 32 USD InStock
77748-088 77748-089
(-)-Huperzine A ≥98%
(-)-Huperzine A
Precaución: for Research Use Only


Formula: C₁₅H₁₈N₂O
MDL Number: MFCD00171439
CAS Number: 102518-79-6
UN: 1544
ADR: 6.1,Ⅱ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty