Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dimethoxy-4-formylphenylboronic acid ≥98%

Supplier: AMBEED, INC
Warning

A1632663-250MG A1632663-100MG A1632663-1G
77625-600EA 88.58 USD InStock
77625-600 77625-596 77625-598
2,5-Dimethoxy-4-formylphenylboronic acid ≥98%
2,5-Dimethoxy-4-formylphenylboronic acid
Precaución: for Research Use Only


Formula: C₉H₁₁BO₅
MW: 209.992 g/mol
MDL Number: MFCD32200303
CAS Number: 2468045-30-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty