Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,3,5-Trimethylcyclohexyl salicylate (cis and trans mixture) ≥98%

Supplier: AMBEED, INC
Warning

A165327-10G A165327-1000G
77615-372EA 14.42 USD InStock
77615-372 77748-100
3,3,5-Trimethylcyclohexyl salicylate (cis and trans mixture) ≥98%
3,3,5-Trimethylcyclohexyl salicylate (cis and trans mixture)
Precaución: for Research Use Only


Formula: C₁₆H₂₂O₃
MDL Number: MFCD00019377
CAS Number: 118-56-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty