Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nɑ-Tosyl-L-arginine methyl ester hydrochloride ≥98%

Supplier: AMBEED, INC
Warning

A169128-100G A169128-25G A169128-5G
77671-394EA 175 USD InStock
77671-394 77671-396 77671-398
Nɑ-Tosyl-L-arginine methyl ester hydrochloride ≥98%
Nalpha-Tosyl-L-arginine methyl ester hydrochloride
Precaución: for Research Use Only


Formula: C₁₄H₂₂N₄O₄S·HCl
MDL Number: MFCD00012578
CAS Number: 1784-03-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty