Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N4-(4-(2-Bromo-4,5-difluorophenoxy)phenyl)-L-asparagine hydrochloride ≥98%

Supplier: AMBEED, INC
Warning

A1720752-10MG A1720752-5MG
77625-786EA 57.68 USD InStock
77625-786 77625-788
N4-(4-(2-Bromo-4,5-difluorophenoxy)phenyl)-L-asparagine hydrochloride ≥98%
N4-(4-(2-Bromo-4,5-difluorophenoxy)phenyl)-L-asparagine hydrochloride
Precaución: for Research Use Only


Formula: C₁₆H₁₄BrClF₂N₂O₄
CAS Number: 2450268-84-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty