Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tigecycline ≥98%

Supplier: AMBEED, INC
Danger

A175151-1G A175151-1MG
77615-464EA 190.55 USD InStock
77615-464 77748-144
Tigecycline ≥98%
Tigecycline
Precaución: for Research Use Only


Formula: C₂₉H₃₉N₅O₈
MDL Number: MFCD00935753
CAS Number: 220620-09-7
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty