Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diacetato{(R)-(-)-5,5'-bis[di(3,5-di-t-butyl-4-methoxyphenyl)phosphino]-4,4'-bi-1,3-benzodioxole}ruthenium(II) ≥98%

Supplier: AMBEED, INC
Warning

A178708-1G A178708-250MG
77746-162EA 316 USD InStock
77746-162 77746-163
Diacetato{(R)-(-)-5,5'-bis[di(3,5-di-t-butyl-4-methoxyphenyl)phosphino]-4,4'-bi-1,3-benzodioxole}ruthenium(II) ≥98%
Diacetato{(R)-(-)-5,5'-bis[di(3,5-di-t-butyl-4-methoxyphenyl)phosphino]-4,4'-bi-1,3-benzodioxole}ruthenium(II)
Precaución: for Research Use Only


Formula: C₇₈H₁₀₆O₁₂P₂Ru
MW: 1398.69 g/mol
MDL Number: MFCD17018804
CAS Number: 1025477-38-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty