Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxy-5-(3,4,5-trimethoxyphenethyl)phenol ≥97%

Supplier: AMBEED, INC
Warning

A184379-100MG A184379-250MG A184379-50MG
77671-602EA 237 USD InStock
77671-602 77671-604 77671-606
2-Methoxy-5-(3,4,5-trimethoxyphenethyl)phenol ≥97%
2-Methoxy-5-(3,4,5-trimethoxyphenethyl)phenol
Precaución: for Research Use Only


Formula: C₁₈H₂₂O₅
MDL Number: MFCD06795132
CAS Number: 95041-90-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty