Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

25-Hydroxycholesterol ≥95%

Supplier: AMBEED, INC
Warning

A185451-10MG A185451-5MG
77746-197EA 58 USD InStock
77746-197 77746-198
25-Hydroxycholesterol ≥95%
25-Hydroxycholesterol
Precaución: for Research Use Only


Formula: C₂₇H₄₆O₂
MDL Number: MFCD00057815
CAS Number: 2140-46-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty