Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Azakenpaullone ≥98%

Supplier: AMBEED, INC
Warning

A192559-10MG A192559-2MG A192559-5MG
77746-227EA 130 USD InStock
77746-227 77746-228 77746-229
1-Azakenpaullone ≥98%
1-Azakenpaullone
Precaución: for Research Use Only


Formula: C₁₅H₁₀BrN₃O
MDL Number: MFCD09037522
CAS Number: 676596-65-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty