Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Betulinic acid ≥98%

Supplier: AMBEED, INC
Warning

A200416-100MG A200416-10G A200416-5G
77671-778EA 8 USD InStock
77671-778 77671-780 77671-782
Betulinic acid ≥98%
Betulinic acid
Precaución: for Research Use Only


Formula: C₃₀H₄₈O₃
MW: 456.71 g/mol
MDL Number: MFCD00009619
CAS Number: 472-15-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty