Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-(2,6-Dioxopiperidin-3-yl)-3-oxoisoindolin-5-yl)methyl (2-fluoro-5-(trifluoromethoxy)phenyl)carbamate ≥98%

Supplier: AMBEED, INC
Warning

A2020160-2MG A2020160-5MG A2020160-10MG
77626-020EA 94.76 USD InStock
77626-020 77626-022 77626-018
(2-(2,6-Dioxopiperidin-3-yl)-3-oxoisoindolin-5-yl)methyl (2-fluoro-5-(trifluoromethoxy)phenyl)carbamate ≥98%
(2-(2,6-Dioxopiperidin-3-yl)-3-oxoisoindolin-5-yl)methyl (2-fluoro-5-(trifluoromethoxy)phenyl)carbamate
Precaución: for Research Use Only


Formula: C₂₂H₁₇F₄N₃O₆
MW: 495.38 g/mol
CAS Number: 2803881-11-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty