Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-2-(trifluoromethyl)benzeneboronic acid pinacol ester ≥97%

Supplier: AMBEED, INC
Warning

A202399-100MG A202399-250MG
77748-274EA 53 USD InStock
77748-274 77748-275
3-Fluoro-2-(trifluoromethyl)benzeneboronic acid pinacol ester ≥97%
3-Fluoro-2-(trifluoromethyl)benzeneboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₃H₁₅BF₄O₂
MW: 290.065 g/mol
MDL Number: MFCD16996330
CAS Number: 1192548-05-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty