Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

A-1331852 ≥98%

Supplier: AMBEED, INC
Warning

A206174-10MG A206174-250MG A206174-50MG
77746-270EA 110 USD InStock
77746-270 77746-271 77746-272
A-1331852 ≥98%
A-1331852
Precaución: for Research Use Only


Formula: C₃₈H₃₈N₆O₃S
MDL Number: MFCD30532662
CAS Number: 1430844-80-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty