Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L-Buthionine sulfoximine ≥98%

Supplier: AMBEED, INC
Warning

A215764-1MG A215764-100MG A215764-50MG
77600-746EA 5.13 USD InStock
77600-746 77667-808 77667-810
L-Buthionine sulfoximine ≥98%
L-Buthionine sulfoximine
Precaución: for Research Use Only


Formula: C₈H₁₈N₂O₃S
MW: 222.303 g/mol
MDL Number: MFCD00067000
CAS Number: 83730-53-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty