Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-Tetrahydro-1H-oxazolo[3,4-a]pyrazin-3(5H)-one hydrochloride ≥97%

Supplier: AMBEED, INC
Warning

A218575-100MG A218575-1G
77746-305EA 13 USD InStock
77746-305 77746-306
(S)-Tetrahydro-1H-oxazolo[3,4-a]pyrazin-3(5H)-one hydrochloride ≥97%
(S)-Tetrahydro-1H-oxazolo[3,4-a]pyrazin-3(5H)-one hydrochloride
Precaución: for Research Use Only


Formula: C₆H₁₁ClN₂O₂
MDL Number: MFCD09878826
CAS Number: 958635-15-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty