Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tazemetostat (EPZ-6438) ≥98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A223202-1MG A223202-10MG A223202-50MG
77612-164EA 6.18 USD InStock
77612-164 77683-642 77683-643
Tazemetostat (EPZ-6438) ≥98%, Ambeed.Inc
Tazemetostat
Precaución: For research use only.


Formula: C₃₄H₄₄N₄O₄
MDL Number: MFCD20527867
CAS Number: 1403254-99-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty