Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methacrylamidoisophthalic acid ≥98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A223586-100MG A223586-1G A223586-250MG
77683-646EA 55 USD InStock
77683-646 77683-647 77683-648
5-Methacrylamidoisophthalic acid ≥98%, Ambeed.Inc
5-Methacrylamidoisophthalic acid
Precaución: For research use only.


Formula: C₁₂H₁₁NO₅
MW: 249.22 g/mol
MDL Number: MFCD00434754
CAS Number: 73912-52-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty