Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1S,2aR,3aS,3bS,4R,5aR,6R,7R,9S,9aS,9bR,11R,11aR,14R)-1-(Furan-3-yl)-4,9,14-trihydroxy-3b,6,11a-trimethyl-10-oxotetradecahydro-1H-6,9a-(methanooxymethano)naphtho[1',2':6,7]indeno[1,7a-b]oxirene-7,11-diyl diacetate ≥98%

Supplier: AMBEED, INC
Danger

A225993-1MG A225993-5MG
77667-876EA 20 USD InStock
77667-876 77667-878
(1S,2aR,3aS,3bS,4R,5aR,6R,7R,9S,9aS,9bR,11R,11aR,14R)-1-(Furan-3-yl)-4,9,14-trihydroxy-3b,6,11a-trimethyl-10-oxotetradecahydro-1H-6,9a-(methanooxymethano)naphtho[1',2':6,7]indeno[1,7a-b]oxirene-7,11-diyl diacetate ≥98%
(1S,2aR,3aS,3bS,4R,5aR,6R,7R,9S,9aS,9bR,11R,11aR,14R)-1-(Furan-3-yl)-4,9,14-trihydroxy-3b,6,11a-trimethyl-10-oxotetradecahydro-1H-6,9a-(methanooxymethano)naphtho[1',2':6,7]indeno[1,7a-b]oxirene-7,11-diyl diacetate
Precaución: for Research Use Only


Formula: C₃₀H₃₈O₁₁
MDL Number: MFCD00210564
CAS Number: 58812-37-6
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty