Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

BMS-582949 ≥98%

Supplier: AMBEED, INC
Danger

A227895-2MG A227895-5MG
77746-337EA 52 USD InStock
77746-337 77746-338
BMS-582949 ≥98%
BMS-582949
Precaución: for Research Use Only


Formula: C₂₂H₂₆N₆O₂
CAS Number: 623152-17-0
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty