Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1R,2S)-2-Fluorocyclopropanecarboxylic acid ≥97%

Supplier: AMBEED, INC
Danger

A237427-10G A237427-250MG
77672-118EA 908 USD InStock
77672-118 77672-120
(1R,2S)-2-Fluorocyclopropanecarboxylic acid ≥97%
(1R,2S)-2-Fluorocyclopropanecarboxylic acid
Precaución: for Research Use Only


Formula: C₄H₅FO₂
MDL Number: MFCD19237658
CAS Number: 167073-08-7
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty