Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

RO5126766 ≥98%

Supplier: AMBEED, INC
Warning

A245684-10MG A245684-5MG
77746-378EA 182 USD InStock
77746-378 77746-379
RO5126766 ≥98%
RO5126766
Precaución: for Research Use Only


Formula: C₂₁H₁₈FN₅O₅S
MDL Number: MFCD27987901
CAS Number: 946128-88-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty