Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Anagrelide ≥98%

Supplier: AMBEED, INC
Warning

A249963-10MG A249963-1MG A249963-5MG
77626-316EA 90.64 USD InStock
77626-316 77626-318 77626-320
Anagrelide ≥98%
Anagrelide
Precaución: for Research Use Only


Formula: C₁₀H₇Cl₂N₃O
MDL Number: MFCD00866794
CAS Number: 68475-42-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty