Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrandine ≥95%

Supplier: AMBEED, INC
Warning

A256378-250MG A256378-5G
77685-729EA 19 USD InStock
77685-729 77685-730
Tetrandine ≥95%
Tetrandine
Precaución: for Research Use Only


Formula: C₃₈H₄₂N₂O₆
MDL Number: MFCD08689909
CAS Number: 518-34-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty