Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nifuratel ≥98%

Supplier: AMBEED, INC
Warning

A260716-10G A260716-25G A260716-5G
77668-122EA 121 USD InStock
77668-122 77668-124 77668-126
Nifuratel ≥98%
Nifuratel


Formula: C₁₀H₁₁N₃O₅S
MW: 285.27 g/mol
MDL Number: MFCD00057257
CAS Number: 4936-47-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty