Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Alfuzosin hydrochloride ≥98%

Supplier: AMBEED, INC
Warning

A272742-250MG A272742-50MG
77626-416EA 60.77 USD InStock
77626-416 77626-418
Alfuzosin hydrochloride ≥98%
Alfuzosin hydrochloride
Precaución: for Research Use Only


Formula: C₁₉H₂₇N₅O₄
MW: 425.91 g/mol
MDL Number: MFCD00879135
CAS Number: 81403-68-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty