Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

CWHM-12 ≥98%

Supplier: AMBEED, INC
Warning

A297047-10MG A297047-2MG A297047-5MG
77746-556EA 148 USD InStock
77746-556 77746-557 77746-558
CWHM-12 ≥98%
CWHM-12
Precaución: for Research Use Only


Formula: C₂₆H₃₂BrN₅O₆
MW: 590.47 g/mol
MDL Number: MFCD28385850
CAS Number: 1564286-55-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty